Anti-BSA 0 Polyclonal Antibody

Supplier: Abnova
PAB0145
10691-564EA 527.69 USD
10691-564
Anti-BSA 0 Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against purified BSA.

Recommended Dilutions: Western Blot: 1:5000; ELISA: 1:5000

Type: Primary
Antigen: BSA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: 0
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR