Anti-EGFR Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10223
10677-848EA 638 USD
10677-848
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of EGFR.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:10000; Immunoprecipitation: 10 ?l

Type: Primary
Antigen: EGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR