Anti-MBD4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0164
10725-058EA 428.44 USD
10725-058
Anti-MBD4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MBD4.

Type: Primary
Antigen: MBD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR