Anti-RBC Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10231
10637-064EA 345.2 USD
10637-064
Anti-RBC Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against native Red Blood Cells (RBC).

Type: Primary
Antigen: RBC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR