Anti-PIAS4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0167
10733-514EA 428.44 USD
10733-514
Anti-PIAS4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PIAS4.

Type: Primary
Antigen: PIAS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR