Anti-ATF2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0410
10680-442EA 346 USD
10680-442
Anti-ATF2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ATF2.

  • Recommended Dilutions: ELISA: 1:1000; Dot Blot: 1:100-500

Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR