Anti-FBXW11 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10237
10714-532EA 638 USD
10714-532
Anti-FBXW11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of FBXW11.

Recommended Dilutions: ELISA: 1:10000-1:100000; Western Blot: 1:200-1:1000

Type: Primary
Antigen: FBXW11
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Bovine, Chicken, Chimpanzee, Dog, Horse, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR