Anti-WRNIP1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10242
10744-694EA 638 USD
10744-694
Anti-WRNIP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of WRNIP1.

Recommended Dilutions: ELISA: 1:10000-1:40000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: WRNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR