Anti-CXCR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0178
10719-302EA 428.44 USD
10719-302
Anti-CXCR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CXCR1.

Type: Primary
Antigen: CXCR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR