Anti-ERICH5 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21762
10704-470EA 428.55 USD
10704-470
Anti-ERICH5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C8orf47.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000

Type: Primary
Antigen: ERICH5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR