Anti-IKBKB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10247
10721-224EA 638 USD
10721-224
Anti-IKBKB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of IKBKB.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: IKBKB
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR