Anti-IL7R Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10248
10719-284EA 786.15 USD
10719-284
Anti-IL7R Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Il7r.

Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:1000-1:10000

Type: Primary
Antigen: IL7R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR