Anti-HIST3H2A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0185
10720-920EA 428.44 USD
10720-920
Anti-HIST3H2A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of HIST3H2A.

Type: Primary
Antigen: HIST3H2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR