Anti-UNG Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0194
10743-850EA 427.45 USD
10743-850
Anti-UNG Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of UNG.

Type: Primary
Antigen: UNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR