Anti-TMEM123 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0204
10741-728EA 527.69 USD
10741-728
Anti-TMEM123 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TMEM123.

Type: Primary
Antigen: TMEM123
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR