Anti-EP300 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0461
10680-544EA 346 USD
10680-544
Anti-EP300 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of EP300.

Recommended Dilutions: ELISA: 1:1000; Dot Blot: 1:500; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: EP300
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR