Anti-FGFR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0472
10680-564EA 346 USD
10680-564
Anti-FGFR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of FGFR1.

Recommended Dilutions: ELISA: 1:1000; Immunohistochemistry: 1:50-100; Western Blot: 1:250-500

Type: Primary
Antigen: FGFR1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR