Anti-NOTCH1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10292
10676-928EA 638 USD
10676-928
Anti-NOTCH1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of NOTCH1.

Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: NOTCH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR