Anti-PRMT5 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0221
10677-078EA 428.44 USD
10677-078
Anti-PRMT5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of PRMT5.

Type: Primary
Antigen: PRMT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR