Anti-CDKN1B Rabbit Polyclonal Antibody

Supplier: Abnova
PAB10300
10707-372EA 638 USD
10707-372
Anti-CDKN1B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant CDKN1B.

Recommended Dilutions: ELISA: 1:10000-1:40000; Western Blot: 1:1000-1:4000

Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR