Anti-CASP8 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0246
10683-420EA 530.32 USD
10683-420
Anti-CASP8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant CASP8.

Type: Primary
Antigen: CASP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Gerbil, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR