Anti-CASP14 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0250
10683-408EA 345.2 USD
10683-408
Anti-CASP14 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant CASP14.

Type: Primary
Antigen: CASP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Dog, Gerbils, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR