Anti-RAD21 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21266
10731-022EA 428.55 USD
10731-022
Anti-RAD21 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant RAD21.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: RAD21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR