Anti-DEDD2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB0255
10683-784EA 329 USD
10683-784
Anti-DEDD2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of DEDD2.

Type: Primary
Antigen: DEDD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR