Anti-RNF34 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11257
10735-620EA 746.62 USD
10735-620
Anti-RNF34 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant RNF34.

Recommended Dilutions: ELISA: 1:4000-1:20000; Western Blot: 1:500-1:3000

Type: Primary
Antigen: RNF34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Orangutan, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR