Anti-MRPL37 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21785
10725-838EA 428.55 USD
10725-838
Anti-MRPL37 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant MRPL37.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Western Blot: 1:250-1:500

Type: Primary
Antigen: MRPL37
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR