Anti-TAF1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11285
10740-444EA 638 USD
10740-444
Anti-TAF1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of TAF1.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:100-1:2000

Type: Primary
Antigen: TAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR