Anti-MANF Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13302
10690-128EA 428 USD
10690-128
Anti-MANF Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ARMET.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: MANF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR