Anti-OTUB Goat Polyclonal Antibody

Supplier: Abnova
PAB11566
10728-202EA 428 USD
10728-202
Anti-OTUB Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of OTUB1.

Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.03-0.1 ?g/ml

Type: Primary
Antigen: OTUB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR