Anti-SERPINB11 Goat Polyclonal Antibody

Supplier: Abnova
PAB11582
10736-856EA 329 USD
10736-856
Anti-SERPINB11 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of SERPINB11.

Recommended Dilutions: ELISA: 1:8000

Type: Primary
Antigen: SERPINB11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR