Anti-Enrofloxacin Monoclonal Antibody [clone: 7F21G1F71]

Supplier: Abnova
MAB2280
10712-066EA 329 USD
10712-066
Anti-Enrofloxacin Monoclonal Antibody [clone: 7F21G1F71]
Antibodies
Mouse monoclonal antibody raised against Enrofloxacin.

Recommended Dilutions: ELISA: 1000-1:2000

Type: Primary
Antigen: Enrofloxacin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 7F21G1F71
Epitope:
Host: IgM
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR