Anti-Ketanserin Monoclonal Antibody [clone: KET101]

Supplier: Abnova
MAB2282
10736-354EA 638 USD
10736-354
Anti-Ketanserin Monoclonal Antibody [clone: KET101]
Antibodies
Mouse monoclonal antibody raised against Ketanserin.

Type: Primary
Antigen: Ketanserin
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: KET101
Epitope:
Host: IgG1
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR