Anti-HMOX1 Mouse Monoclonal Antibody [clone: GTS-1]

Supplier: Abnova
MAB2287
10717-648EA 638 USD
10717-648
Anti-HMOX1 Mouse Monoclonal Antibody [clone: GTS-1]
Antibodies
Mouse monoclonal antibody raised against Hmox1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 5-10 ?g/ml; Immunohistochemistry (Frozen sections): 5-10 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5-10 ?g/ml

Type: Primary
Antigen: HMOX1
Clonality: Monoclonal
Clone: GTS-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR