Anti-STRN3 Mouse Monoclonal Antibody [clone: S68]

Supplier: Abnova
MAB2291
10739-954EA 530.32 USD
10739-954
Anti-STRN3 Mouse Monoclonal Antibody [clone: S68]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of STRN3.

Recommended Dilutions: Immunofluorescence: 1:5000; Western Blot: 1:250-1:2000

Type: Primary
Antigen: STRN3
Clonality: Monoclonal
Clone: S68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR