Anti-SLC9A3 Mouse Monoclonal Antibody [clone: 14D5]

Supplier: Abnova
MAB2294
10675-324EA 530.14 USD
10675-324
Anti-SLC9A3 Mouse Monoclonal Antibody [clone: 14D5]
Antibodies
Mouse monoclonal antibody raised against synthetic phosphopeptide of Slc9a3.

Recommended Dilutions: Immunohistochemistry: 1:50-1:1000; Immunofluorescence: 1:50-1:1000; Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SLC9A3
Clonality: Monoclonal
Clone: 14D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Opossum, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR