Anti-CDK12 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2066
10678-480EA 329 USD
10678-480
Anti-CDK12 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CRKRS.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: CDK12
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR