Anti-NF1 Mouse Monoclonal Antibody [clone: McNFn27a]

Supplier: Abnova
MAB2305
10676-910EA 530.32 USD
10676-910
Anti-NF1 Mouse Monoclonal Antibody [clone: McNFn27a]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of NF1.

Recommended Dilutions: Western Blot: 2-5 ?g/ml

Type: Primary
Antigen: NF1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: McNFn27a
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Guinea Pig, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR