Anti-ABCA4 Goat Polyclonal Antibody

Supplier: Abnova
PAB11617
10687-590EA 329 USD
10687-590
Anti-ABCA4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of ABCA4.

Recommended Dilutions: ELISA: 1:32000

Type: Primary
Antigen: ABCA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR