Anti-DYRK1A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2067
10678-610EA 329 USD
10678-610
Anti-DYRK1A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of DYRK1A.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-100; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: DYRK1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR