Anti-T. gondii Mouse Monoclonal Antibody [clone: TP3]

Supplier: Abnova
MAB3200
10677-412EA 329 USD
10677-412
Anti-T. gondii Mouse Monoclonal Antibody [clone: TP3]
Antibodies
Mouse monoclonal antibody raised against Tachizoites.

Type: Primary
Antigen: T. gondii
Clonality: Monoclonal
Clone: TP3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Protozoa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR