Anti-MMP16 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12293
10732-786EA 428.44 USD
10732-786
Anti-MMP16 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MMP16.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 0.5-1 ?g/ml; Immunohistochemistry (Frozen sections): 0.5-1 ?g/ml

Type: Primary
Antigen: MMP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR