Anti-PPP1R15A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21284
10729-902EA 428.55 USD
10729-902
Anti-PPP1R15A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PPP1R15A.

Type: Primary
Antigen: PPP1R15A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR