Anti-PTPRC Rat Monoclonal Antibody [clone: C363.16A]

Supplier: Abnova
MAB3233
10735-588EA 346 USD
10735-588
Anti-PTPRC Rat Monoclonal Antibody [clone: C363.16A]
Antibodies
Rat monoclonal antibody raised against Ptprc exon B-dependent form.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: C363.16A
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR