Anti-CD86 Rat Monoclonal Antibody [clone: GL1]

Supplier: Abnova
MAB3238
10734-776EA 346 USD
10734-776
Anti-CD86 Rat Monoclonal Antibody [clone: GL1]
Antibodies
Rat monoclonal antibody raised against Cd86.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR