Anti-CXCR4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12323
10709-614EA 428.44 USD
10709-614
Anti-CXCR4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CXCR4.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2 ?g/ml

Type: Primary
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR