Anti-ROCK2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12643
10675-300EA 346 USD
10675-300
Anti-ROCK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ROCK2.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml; Immunohistochemistry: 0.5-2 ?g/ml

Type: Primary
Antigen: ROCK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR