Anti-CHIC2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB21837
10710-842EA 428.55 USD
10710-842
Anti-CHIC2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant CHIC2.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: CHIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR