Anti-Cytomegalovirus glycoprotein B Mouse Monoclonal Antibody [clone: 1-M-12]

Supplier: Abnova
MAB3969
10708-358EA 638.45 USD
10708-358
Anti-Cytomegalovirus glycoprotein B Mouse Monoclonal Antibody [clone: 1-M-12]
Antibodies
Mouse monoclonal antibody raised against Cytomegalovirus glycoprotein B.

Recommended Dilutions: immunofluorescence(1:1000; ELISA: 1:10000

Type: Primary
Antigen: Cytomegalovirus glycoprotein B
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 1-M-12
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR