Anti-SLITRK6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13336
10738-692EA 428 USD
10738-692
Anti-SLITRK6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of SLITRK6.

  • Recommended Dilutions: Western Blot: 0.5-1 μg/ml

Type: Primary
Antigen: SLITRK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR