Anti-THC Monoclonal Antibody [clone: C29]

Supplier: Abnova
MAB4736
10628-874EA 428.44 USD
10628-874
Anti-THC Monoclonal Antibody [clone: C29]
Antibodies
Mouse monoclonal antibody raised against THC.

Type: Primary
Antigen: THC
Clonality: Monoclonal
Clone: C29
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR